Gene Transcripts

Gene Symbol
GATB
Entrez Gene ID
5188
Gene Name
glutamyl-tRNA amidotransferase subunit B
Gene Aliases
HSPC199, PET112, PET112L
Chromosome Location
Chr. 4: 151669773 - 151761023 on Build GRCh38
UniGene ID
Hs.119316
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004564.2 NP_004555.1 1, 2 216
XM_005263063.3 XP_005263120.1 1, 2 178
GenBank mRNA AB019410.2 - 1, 2 160
AF026851.1 - 1, 2 187
AF151033.1 - 1, 2 184
AK222887.1 - 1, 2 178
AK225241.1 - 1, 2 178
AK303461.1 - 1, 2 163
AK312957.1 - 1, 2 178
BC130348.1 - 1, 2 226 40146379
BC136547.1 - 1, 2 226 9020537
BP348031.1 - 1, 2 216

View Full Product Details