Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256045.1 | NP_001242974.1 | 10 | 1587 | |
NM_006219.2 | NP_006210.1 | 19 | 2721 | ||
XM_005247530.2 | XP_005247587.1 | 21 | 3070 | ||
XM_006713659.3 | XP_006713722.1 | 20 | 2941 | ||
XM_011512895.2 | XP_011511197.1 | 22 | 3066 | ||
XM_011512896.1 | XP_011511198.1 | 17 | 2335 | ||
XM_017006619.1 | XP_016862108.1 | 20 | 2744 | ||
XM_017006620.1 | XP_016862109.1 | 19 | 2491 | ||
XM_017006621.1 | XP_016862110.1 | 18 | 2440 | ||
GenBank mRNA | AK293754.1 | - | 10 | 1587 | |
AK302937.1 | - | 12 | 1600 | ||
BC003393.1 | - | 4 | 354 | ||
BC114432.1 | - | 19 | 2709 | 40008544 | |
CR749357.1 | - | 12 | 1306 | ||
S67334.1 | - | 19 | 2705 |