Gene Transcripts

Gene Symbol
IFRD1
Entrez Gene ID
3475
Gene Name
interferon related developmental regulator 1
Gene Aliases
PC4, TIS7
Chromosome Location
Chr. 7: 112423144 - 112477203 on Build GRCh38
UniGene ID
Hs.744747
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007245.2 NP_001007246.1 8 1236
NM_001197079.1 NP_001184008.1 7 1126
NM_001197080.1 NP_001184009.1 7 961
NM_001550.3 NP_001541.2 7 1027
NR_120333.1 6 789
XM_011516142.2 XP_011514444.1 7 1022
XM_011516143.2 XP_011514445.1 7 1207
XM_017012151.1 XP_016867640.1 7 1274
GenBank mRNA AK297509.1 - 6 614
AK298894.1 - 7 767
AK308066.1 - 8 1211
AK313211.1 - 8 1236
AK315967.1 - 7 959
BC001272.1 - 7 980 3458309
BT019889.1 - 7 766
BT019890.1 - 7 766

View Full Product Details