Gene Transcripts
- Gene Symbol
- KCND1
- Entrez Gene ID
- 3750
- Gene Name
- potassium voltage-gated channel subfamily D member 1
- Gene Aliases
- KV4.1
- Chromosome Location
- Chr. X: 48960983 - 48972099 on Build GRCh38
- UniGene ID
- Hs.55276
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004979.5 |
NP_004970.3
|
4, 5 |
3047 |
|
XM_011543910.2 |
XP_011542212.1
|
5, 6 |
1955 |
|
XM_017029508.1 |
XP_016884997.1
|
5, 6 |
1786 |
|
XM_017029509.1 |
XP_016884998.1
|
5, 6 |
1783 |
|
GenBank mRNA |
AB021865.1 |
- |
4, 5 |
2772 |
|
AF166003.1 |
- |
4, 5 |
1524 |
|
AJ005898.1 |
- |
4, 5 |
1474 |
|
AK094431.1 |
- |
3, 4 |
1633 |
|
AK315092.1 |
- |
4, 5 |
1958 |
|
BC045659.1 |
- |
4, 5 |
3045 |
5271042 |
View Full Product Details