Gene Transcripts

Gene Symbol
CASQ2
Entrez Gene ID
845
Gene Name
calsequestrin 2
Gene Aliases
PDIB2
Chromosome Location
Chr. 1: 115700003 - 115768805 on Build GRCh38
UniGene ID
Hs.57975
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001232.3 NP_001223.2 11 2508
GenBank mRNA AK129831.1 - 3 1363
AK129891.1 - 11 2485
BC022288.1 - 11 2416 4289141
D55655.1 - 11 2353

View Full Product Details