Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001303110.1 | NP_001290039.1 | 21, 22 | 1410 | |
NM_001845.5 | NP_001836.3 | 21, 22 | 1410 | ||
XM_011521048.2 | XP_011519350.1 | 21, 22 | 9342 | ||
GenBank mRNA | AB209646.1 | - | 21, 22 | 1395 | |
BC142626.1 | - | 21, 22 | 1384 | ||
BC151220.1 | - | 21, 22 | 1394 | 40148649 | |
X05561.1 | - | 21, 22 | 1402 | ||
Y00706.1 | - | 19, 20 | 1145 |