Gene Transcripts

Gene Symbol
DPT
Entrez Gene ID
1805
Gene Name
dermatopontin
Gene Aliases
TRAMP
Chromosome Location
Chr. 1: 168695457 - 168729204 on Build GRCh38
UniGene ID
Hs.80552
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001937.4 NP_001928.2 4 1058
GenBank mRNA AL049455.1 - 1 217
BC033736.1 - 4 1044 5176855

View Full Product Details