Gene Transcripts

Gene Symbol
F2RL2
Entrez Gene ID
2151
Gene Name
coagulation factor II thrombin receptor like 2
Gene Aliases
PAR-3, PAR3
Chromosome Location
Chr. 5: 76615482 - 76623434 on Build GRCh38
UniGene ID
Hs.42502
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256566.1 NP_001243495.1 2 1866
NM_004101.3 NP_004092.1 2 2017
GenBank mRNA BX537386.1 - 2 1996

View Full Product Details