Gene Transcripts

Gene Symbol
CSNK1G3
Entrez Gene ID
1456
Gene Name
casein kinase 1 gamma 3
Gene Aliases
CKI-gamma 3, CSNK1G3L
Chromosome Location
Chr. 5: 123512099 - 123617045 on Build GRCh38
UniGene ID
Hs.129206
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031812.3 NP_001026982.1 10, 11 1813
NM_001044723.2 NP_001038188.1 10, 11 1437
NM_001270572.1 NP_001257501.1 9, 10 1344
NM_001270573.1 NP_001257502.1 9, 10 1388
NM_001270574.1 NP_001257503.1 7, 8 1280
NM_004384.4 NP_004375.2 10, 11 1437
XM_005271891.1 XP_005271948.1 11, 12 1912
XM_005271892.2 XP_005271949.1 11, 12 1909
XM_005271894.2 XP_005271951.1 11, 12 1909
XM_005271895.2 XP_005271952.1 10, 11 1816
XM_005271896.1 XP_005271953.1 10, 11 1816
XM_017009057.1 XP_016864546.1 12, 13 1987
XM_017009058.1 XP_016864547.1 14, 15 2196
XM_017009059.1 XP_016864548.1 13, 14 2055
XM_017009060.1 XP_016864549.1 13, 14 2111
XM_017009061.1 XP_016864550.1 12, 13 1984
XM_017009062.1 XP_016864551.1 12, 13 1543
XM_017009063.1 XP_016864552.1 12, 13 1987
XM_017009064.1 XP_016864553.1 11, 12 1891
XM_017009065.1 XP_016864554.1 11, 12 1888
XM_017009066.1 XP_016864555.1 12, 13 1956
XM_017009067.1 XP_016864556.1 11, 12 1891
XM_017009068.1 XP_016864557.1 11, 12 1888
XM_017009069.1 XP_016864558.1 11, 12 1884
XM_017009070.1 XP_016864559.1 10, 11 1813
GenBank mRNA AF049089.1 - 10, 11 1220
AF049090.1 - 10, 11 1220
AK289405.1 - 9, 10 1289
AK299739.1 - 9, 10 1227
AK316019.1 - 7, 8 1119
BC047567.1 - 10, 11 1465 4817058

View Full Product Details