Gene Transcripts

Gene Symbol
C6
Entrez Gene ID
729
Gene Name
complement component 6
Gene Aliases
Chromosome Location
Chr. 5: 41142116 - 41261486 on Build GRCh38
UniGene ID
Hs.481992
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000065.3 NP_000056.2 3 447
NM_001115131.2 NP_001108603.2 3 474
XM_005248357.2 XP_005248414.1 3 522
XM_006714496.3 XP_006714559.1 3 522
XM_011514114.2 XP_011512416.1 3 522
XM_011514115.2 XP_011512417.1 3 443
XM_011514116.2 XP_011512418.1 4 559
XM_011514117.2 XP_011512419.1 4 601
XM_011514118.2 XP_011512420.1 3 339
XM_011514119.2 XP_011512421.1 3 522
XM_017009818.1 XP_016865307.1 4 582
GenBank mRNA AB126592.1 - 2 209
AK292509.1 - 3 345
AK313673.1 - 3 366
BC035723.1 - 3 474 5591446
DB066614.1 - 3 447
DB184605.1 - 3 419
J04506.1 - 3 275
J05024.1 - 3 266
J05064.1 - 3 364

View Full Product Details