Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005892.3 | NP_005883.2 | 17 | 2322 | |
XM_006722062.3 | XP_006722125.1 | 18 | 2537 | ||
XM_006722063.3 | XP_006722126.1 | 17 | 2517 | ||
XM_006722064.3 | XP_006722127.1 | 18 | 2507 | ||
XM_006722065.3 | XP_006722128.1 | 17 | 2487 | ||
XM_006722066.3 | XP_006722129.1 | 18 | 2533 | ||
XM_006722069.2 | XP_006722132.1 | 18 | 2536 | ||
XM_006722070.3 | XP_006722133.1 | 17 | 2513 | ||
XM_011525179.2 | XP_011523481.1 | 18 | 2537 | ||
XM_011525180.2 | XP_011523482.1 | 18 | 2229 | ||
XM_011525182.2 | XP_011523484.1 | 18 | 2534 | ||
GenBank mRNA | AF432213.1 | - | 14 | 1796 | |
AJ008112.1 | - | 2 | 238 | ||
AK131091.1 | - | 9 | 2859 | ||
AY278319.1 | - | 17 | 2322 | ||
BC001710.1 | - | 2 | 226 | ||
BC021906.1 | - | 5 | 830 | ||
BC073988.1 | - | 5 | 870 | ||
BC111564.1 | - | 10 | 1135 | 40008718 | |
CR456759.1 | - | 2 | 199 | ||
FJ534522.1 | - | 1 | 121 |