Gene Transcripts

Gene Symbol
CIB3
Entrez Gene ID
117286
Gene Name
calcium and integrin binding family member 3
Gene Aliases
KIP3
Chromosome Location
Chr. 19: 16161368 - 16173525 on Build GRCh38
UniGene ID
Hs.255432
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300922.1 NP_001287851.1 3 271
NM_054113.3 NP_473454.1 5 418
GenBank mRNA AB050868.1 - 5 368
BC069320.1 - 3 108
BC069428.1 - 3 221 7262131
BC069524.1 - 5 349
BC112200.1 - 5 418 8327668
BC113591.1 - 5 418 8322643

View Full Product Details