Gene Transcripts

Gene Symbol
FBXL13
Entrez Gene ID
222235
Gene Name
F-box and leucine rich repeat protein 13
Gene Aliases
DRC6, Fbl13
Chromosome Location
Chr. 7: 102699226 - 103074841 on Build GRCh38
UniGene ID
Hs.660029
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111038.1 NP_001104508.1 14 1611
NM_001287150.1 NP_001274079.1 14 1611
NM_145032.3 NP_659469.3 14 1788
NR_105043.1 15 1702
XM_005250205.3 XP_005250262.1 15 1694
XM_005250207.3 XP_005250264.1 14 1595
XM_005250208.3 XP_005250265.1 15 1694
XM_005250209.2 XP_005250266.1 14 1592
XM_006715898.2 XP_006715961.1 9 1039
XM_011515928.2 XP_011514230.1 15 1694
XM_011515929.2 XP_011514231.1 15 1694
XM_011515930.2 XP_011514232.1 14 1658
XM_011515932.2 XP_011514234.1 15 1694
XM_017011850.1 XP_016867339.1 15 1638
XM_017011851.1 XP_016867340.1 15 1694
XM_017011852.1 XP_016867341.1 15 1844
XM_017011853.1 XP_016867342.1 14 1644
GenBank mRNA AK095387.1 - 9 1011
AK097537.1 - 14 1788
AL713709.1 - 5 510
AY359239.1 - 14 1495
BC020572.2 - 8 865
BC026121.1 - 15 1705 4826546
BC031285.1 - 14 1603 5298780

View Full Product Details