Gene Transcripts

Gene Symbol
OTOS
Entrez Gene ID
150677
Gene Name
otospiralin
Gene Aliases
OTOSP
Chromosome Location
Chr. 2: 240138942 - 240141373 on Build GRCh38
UniGene ID
Hs.148586
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_148961.3 NP_683764.1 4 546
XM_017003409.1 XP_016858898.1 4 500
XM_017003410.1 XP_016858899.1 4 1181

View Full Product Details