Gene Transcripts

Gene Symbol
ZZEF1
Entrez Gene ID
23140
Gene Name
zinc finger ZZ-type and EF-hand domain containing 1
Gene Aliases
ZZZ4
Chromosome Location
Chr. 17: 4004445 - 4143020 on Build GRCh38
UniGene ID
Hs.277624
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015113.3 NP_055928.3 5, 6 1134
XM_005256561.2 XP_005256618.1 5, 6 1195
XM_011523759.2 XP_011522061.1 5, 6 1195
XM_017024382.1 XP_016879871.1 5, 6 1195
XM_017024383.1 XP_016879872.1 5, 6 1195
GenBank mRNA AB007859.4 - 5, 6 1134
AK127482.1 - 5, 6 1195
BC151836.1 - 5, 6 1134 8860131

View Full Product Details