Gene Transcripts

Gene Symbol
MGRN1
Entrez Gene ID
23295
Gene Name
mahogunin ring finger 1
Gene Aliases
RNF156
Chromosome Location
Chr. 16: 4616493 - 4690974 on Build GRCh38
UniGene ID
Hs.526494
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142289.2 NP_001135761.2 3, 4 441
NM_001142290.2 NP_001135762.1 3, 4 441
NM_001142291.2 NP_001135763.2 3, 4 441
NM_015246.3 NP_056061.1 3, 4 441
NR_102267.1 3, 4 441
XM_005255217.1 XP_005255274.1 3, 4 441
XM_005255218.1 XP_005255275.1 3, 4 441
XM_005255219.3 XP_005255276.1 3, 4 441
XM_005255220.3 XP_005255277.1 3, 4 441
XM_005255221.3 XP_005255278.1 3, 4 441
XM_011522441.1 XP_011520743.1 3, 4 299
XM_017023091.1 XP_016878580.1 3, 4 441
XM_017023092.1 XP_016878581.1 3, 4 441
XM_017023093.1 XP_016878582.1 3, 4 299
GenBank mRNA AB011116.1 - 3, 4 397
AK299052.1 - 3, 4 423
BC050389.1 - 3, 4 413 5755971
CN314787.1 - 3, 4 402
DA118421.1 - 3, 4 440
DA590130.1 - 3, 4 441
EF471397.2 - 3, 4 304
EF471398.2 - 3, 4 304

View Full Product Details