Gene Transcripts

Gene Symbol
MGRN1
Entrez Gene ID
23295
Gene Name
mahogunin ring finger 1
Gene Aliases
RNF156
Chromosome Location
Chr. 16: 4616493 - 4690974 on Build GRCh38
UniGene ID
Hs.526494
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142289.2 NP_001135761.2 16 2249
NM_001142290.2 NP_001135762.1 17 4852
NM_001142291.2 NP_001135763.2 16 4786
NM_015246.3 NP_056061.1 17 2315
NR_102267.1 15 2139
XM_005255217.1 XP_005255274.1 17 2318
XM_005255218.1 XP_005255275.1 16 2252
XM_011522441.1 XP_011520743.1 17 2176
XM_011522442.2 XP_011520744.1 9 1630
GenBank mRNA AB011116.1 - 17 4832
BC007627.1 - 1 1437
BC050389.1 - 17 2287 5755971

View Full Product Details