Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001162536.2 | NP_001156008.1 | 3 | 3775 | |
NM_019610.5 | NP_062556.2 | 2 | 3675 | ||
GenBank mRNA | AL832554.1 | - | 2 | 3528 | |
AL832785.1 | - | 1 | 2616 | ||
BC012942.1 | - | 2 | 3334 | 3875565 |