Gene Transcripts

Gene Symbol
LZTS2
Entrez Gene ID
84445
Gene Name
leucine zipper tumor suppressor 2
Gene Aliases
LAPSER1
Chromosome Location
Chr. 10: 100997065 - 101007836 on Build GRCh38
UniGene ID
Hs.523221
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318099.1 NP_001305028.1 5 2348
NM_001318100.1 NP_001305029.1 5 2425
NM_001318101.1 NP_001305030.1 3 1626
NM_032429.3 NP_115805.1 5 2455
XM_005270222.4 XP_005270279.1 5 2687
XM_017016780.1 XP_016872269.1 4 4816
XM_017016781.1 XP_016872270.1 4 1796
GenBank mRNA AL834338.1 - 3 1831
BC005855.2 - 2 1159
BC006212.2 - 5 2313 3161855

View Full Product Details