Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317958.1 | NP_001304887.1 | 9 | 1347 | |
NM_001317959.1 | NP_001304888.1 | 11 | 1733 | ||
NM_152572.2 | NP_689785.1 | 9 | 1340 | ||
XM_005272169.2 | XP_005272226.1 | 9 | 879 | ||
XM_006716965.2 | XP_006717028.1 | 7 | 652 | ||
XM_011518277.2 | XP_011516579.1 | 9 | 879 | ||
XM_011518278.2 | XP_011516580.1 | 8 | 720 | ||
XM_017014308.1 | XP_016869797.1 | 3 | 402 | ||
GenBank mRNA | AK057266.1 | - | 9 | 813 | |
AK093446.1 | - | 9 | 855 | ||
AK292186.1 | - | 9 | 825 | ||
BC034776.1 | - | 9 | 837 | 5175757 | |
BC050576.1 | - | 9 | 1340 | 5744517 |