Gene Transcripts

Gene Symbol
KRT75
Entrez Gene ID
9119
Gene Name
keratin 75
Gene Aliases
K6HF, KB18, PFB
Chromosome Location
Chr. 12: 52424070 - 52434326 on Build GRCh38
UniGene ID
Hs.697046
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004693.2 NP_004684.2 2 689
GenBank mRNA AK225962.1 - 2 689
AK298960.1 - 2 888
Y17282.1 - 2 685

View Full Product Details