Gene Transcripts

Gene Symbol
Pitx2
Entrez Gene ID
18741
Gene Name
paired-like homeodomain transcription factor 2
Gene Aliases
9430085M16Rik, Brx1, Brx1b, Munc30, Otlx2, Ptx2, Rieg
Chromosome Location
Chr. 3: 129199878 - 129219594 on Build GRCm38
UniGene ID
Mm.246804
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042502.2 NP_001035967.1 3 1996
NM_001042504.2 NP_001035969.1 4 1487
NM_001286942.1 NP_001273871.1 5 1586
NM_001287048.1 NP_001273977.1 3 1996
NM_011098.4 NP_035228.2 5 1625
GenBank mRNA AF048723.1 - 4 1423
AF048724.1 - 3 1956
AF201091.1 - 3 1323
BC075660.1 - 3 1661 30606653
U70132.1 - 2 1146
U80010.1 - 5 1560
U80011.1 - 4 1440
U80036.1 - 4 1458

View Full Product Details