Gene Transcripts

Gene Symbol
NPHS1
Entrez Gene ID
4868
Gene Name
NPHS1, nephrin
Gene Aliases
CNF, NPHN, nephrin
Chromosome Location
Chr. 19: 35825372 - 35851993 on Build GRCh38
UniGene ID
Hs.122186
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004646.3 NP_004637.1 21 3000
GenBank mRNA AF035835.1 - 21 2844
AF190637.1 - 21 2844
EU642886.1 - 21 2844
EU642887.1 - 21 2843

View Full Product Details