Gene Transcripts

Gene Symbol
St6galnac1
Entrez Gene ID
20445
Gene Name
ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 1
Gene Aliases
Siat7a
Chromosome Location
Chr. 11: 116765025 - 116775507 on Build GRCm38
UniGene ID
Mm.436887
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011371.2 NP_035501.2 2 695
GenBank mRNA BC141095.1 - 678
BC145373.1 - 678 9053931
BC145374.1 - 2 678 9053932
Y11274.1 - 2 695

View Full Product Details