Gene Transcripts

Gene Symbol
Rab4b
Entrez Gene ID
19342
Gene Name
RAB4B, member RAS oncogene family
Gene Aliases
1500031G17Rik
Chromosome Location
Chr. 7: 27168424 - 27178901 on Build GRCm38
UniGene ID
Mm.482114
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029391.2 NP_083667.1 7 705
GenBank mRNA AB232592.1 - 7 564
AF408432.1 - 7 677
AK005314.1 - 7 705
AK140513.1 - 6 902
AK170552.1 - 7 719
AK219531.1 - 3 254
BC007147.1 - 7 649 3600193
BC096654.1 - 7 688 5056766

View Full Product Details