Gene Transcripts
- Gene Symbol
- Mllt6
- Entrez Gene ID
- 246198
- Gene Name
- myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 6
- Gene Aliases
- AI315037, Af17
- Chromosome Location
- Chr. 11: 97663412 - 97685461 on Build GRCm38
- UniGene ID
- Mm.23685
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY050217.1 |
- |
|
4471 |
|
View Full Product Details