Gene Transcripts

Gene Symbol
Emilin2
Entrez Gene ID
246707
Gene Name
elastin microfibril interfacer 2
Gene Aliases
FOAP-10
Chromosome Location
Chr. 17: 71252172 - 71311639 on Build GRCm38
UniGene ID
Mm.23462
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145158.3 NP_660140.1 3 522
GenBank mRNA AF468645.1 - 3 473
AK154230.1 - 3 492
AK155037.1 - 3 500
AK155306.1 - 3 516
AK155366.1 - 3 502
AK155554.1 - 3 518
AK155797.1 - 3 519
AK155878.1 - 3 520
AK155879.1 - 3 516
AK155909.1 - 3 522
AK155913.1 - 3 492
AK155918.1 - 3 549
AK170081.1 - 3 519
AK170288.1 - 3 492
AK170391.1 - 3 491
AK170801.1 - 3 523
AK170822.1 - 3 544
AK170953.1 - 3 538
AK171015.1 - 3 544
AK171020.1 - 3 510
AK171303.1 - 3 494
AK171309.1 - 3 519
AK171579.1 - 3 492
AK171592.1 - 3 492
AK171616.1 - 3 492
BC053753.1 - 3 806 6510220
GQ872457.1 - 3 417
GQ872460.1 - 3 417

View Full Product Details