Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145589.2 | NP_663564.2 | 8 | 1709 | |
GenBank mRNA | AK133884.1 | - | 8 | 1673 | |
AK139559.1 | - | 9 | 1484 | ||
AK145816.1 | - | 9 | 1489 | ||
AK151203.1 | - | 9 | 1541 | ||
AK182875.1 | - | 2 | 147 | ||
AK207496.1 | - | 1 | 66 | ||
BC055033.1 | - | 8 | 1709 | 6310272 |