Gene Transcripts
- Gene Symbol
- Cables2
- Entrez Gene ID
- 252966
- Gene Name
- CDK5 and Abl enzyme substrate 2
- Gene Aliases
- ik3-2
- Chromosome Location
- Chr. 2: 180258539 - 180273490 on Build GRCm38
- UniGene ID
- Mm.386884
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145851.2 |
NP_665850.2
|
3, 4 |
536 |
|
GenBank mRNA |
AK050121.1 |
- |
3, 4 |
536 |
|
View Full Product Details