Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_148939.2 | NP_683741.1 | 3 | 527 | |
GenBank mRNA | AJ315553.1 | - | 3 | 527 | |
AJ315554.1 | - | 2 | 623 | ||
AK151902.1 | - | 2 | 564 | ||
BC131964.1 | - | 3 | 555 | 40130241 | |
BC131966.1 | - | 3 | 555 | 40130243 | |
BC145150.1 | - | 2 | 651 | ||
HE864438.1 | - | 3 | 543 | ||
HE864439.1 | - | 2 | 639 |