Gene Transcripts

Gene Symbol
Ttc34
Entrez Gene ID
242800
Gene Name
tetratricopeptide repeat domain 34
Gene Aliases
4932418L02, B230396O12Rik
Chromosome Location
Chr. 4: 154833443 - 154867127 on Build GRCm38
UniGene ID
Mm.72944
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172878.3 NP_766466.3 2 274
GenBank mRNA AK030055.1 - 2 248
BC068299.1 - 126 30094917
CJ183638.1 - 2 277

View Full Product Details