Gene Transcripts

Gene Symbol
Clec1a
Entrez Gene ID
243653
Gene Name
C-type lectin domain family 1, member a
Gene Aliases
5930406N14Rik
Chromosome Location
Chr. 6: 129423358 - 129452000 on Build GRCm38
UniGene ID
Mm.131289
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175526.3 NP_780735.2 6 2232
GenBank mRNA AK031121.1 - 6 2052
AK036846.1 - 6 2037

View Full Product Details