Gene Transcripts

Gene Symbol
Relt
Entrez Gene ID
320100
Gene Name
RELT tumor necrosis factor receptor
Gene Aliases
E430021K24Rik, Tnfrsf19l
Chromosome Location
Chr. 7: 100845847 - 100863480 on Build GRCm38
UniGene ID
Mm.40336
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177073.6 NP_796047.2 10 1352
GenBank mRNA AK042363.1 - 8 1185
AK049073.1 - 10 1353
AK088621.1 - 10 1335
BC100310.1 - 10 1342 30839974
BC131929.1 - 10 1263 40130206
BC131931.1 - 10 1263 40130208
BC145200.1 - 10 1263 9053751

View Full Product Details