Gene Transcripts

Gene Symbol
MRPL33
Entrez Gene ID
9553
Gene Name
mitochondrial ribosomal protein L33
Gene Aliases
C2orf1, L33mt, MRP-L33, RPL33L
Chromosome Location
Chr. 2: 27771717 - 27779741 on Build GRCh38
UniGene ID
Hs.515879
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004891.3 NP_004882.1 4 359
NM_145330.2 NP_663303.1 3 252
GenBank mRNA AF047440.1 - 4 333
BC009475.1 - 4 311 4049053
BC020834.1 - 4 346 4775045
BC087846.1 - 3 216
BC090047.1 - 3 252 30393466
BI601959.1 - 4 362

View Full Product Details