Gene Transcripts

Gene Symbol
Pkd1l3
Entrez Gene ID
244646
Gene Name
polycystic kidney disease 1 like 3
Gene Aliases
Chromosome Location
Chr. 8: 109613994 - 109673526 on Build GRCm38
UniGene ID
Mm.436810
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039700.2 NP_001034789.2 1 159
NM_001286454.1 NP_001273383.1 1 159
NM_181544.2 NP_853522.2 1 159
GenBank mRNA AB290926.1 - 1 159
AY164486.1 - 1 159
DQ382344.1 - 1 159
DQ382345.1 - 1 159
DQ382346.1 - 1 159

View Full Product Details