Gene Transcripts

Gene Symbol
Man2b2
Entrez Gene ID
17160
Gene Name
mannosidase 2, alpha B2
Gene Aliases
mKIAA0935
Chromosome Location
Chr. 5: 36806813 - 36830649 on Build GRCm38
UniGene ID
Mm.761
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008550.2 NP_032576.2 11 1905
GenBank mRNA AB006458.1 - 11 1883
AK004900.1 - 11 1859
AK040388.1 - 11 1866
AK075649.1 - 3 570
AK165376.1 - 11 1898
BC066211.1 - 11 1856 30101816

View Full Product Details