Gene Transcripts

Gene Symbol
Nab1
Entrez Gene ID
17936
Gene Name
Ngfi-A binding protein 1
Gene Aliases
Chromosome Location
Chr. 1: 52455849 - 52500448 on Build GRCm38
UniGene ID
Mm.25903
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008667.3 NP_032693.2 8 2916
GenBank mRNA AK018122.1 - 2 1328
BC016886.1 - 8 2839 4236565

View Full Product Details