Gene Transcripts

Gene Symbol
Neo1
Entrez Gene ID
18007
Gene Name
neogenin
Gene Aliases
2610028H22Rik, AI327052, D930014N22Rik, Igdcc2
Chromosome Location
Chr. 9: 58874679 - 59036957 on Build GRCm38
UniGene ID
Mm.42249
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042752.1 NP_001036217.1 2 717
NM_008684.2 NP_032710.2 2 717
GenBank mRNA AK084609.1 - 2 706
BC054540.1 - 2 701 6406007
Y09535.1 - 2 584

View Full Product Details