Gene Transcripts

Gene Symbol
Pdgfa
Entrez Gene ID
18590
Gene Name
platelet derived growth factor, alpha
Gene Aliases
Chromosome Location
Chr. 5: 138976381 - 138999916 on Build GRCm38
UniGene ID
Mm.2675
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008808.3 NP_032834.1 4 489
GenBank mRNA AK017169.1 - 2 74
AY324648.1 - 4 478
BC003817.1 - 4 489 3495629
M29464.1 - 4 369

View Full Product Details