Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011044.2 | NP_035174.1 | 3, 4 | 554 | |
GenBank mRNA | AK028046.1 | - | 3, 4 | 555 | |
AK030327.1 | - | 3, 4 | 554 | ||
AK133496.1 | - | 3, 4 | 555 | ||
AK149525.1 | - | 3, 4 | 555 | ||
BC028342.1 | - | 3, 4 | 556 | ||
BC037629.1 | - | 3, 4 | 549 | 4973187 |