Gene Transcripts

Gene Symbol
Kcna6
Entrez Gene ID
16494
Gene Name
potassium voltage-gated channel, shaker-related, subfamily, member 6
Gene Aliases
Kv1.6, MK1.6
Chromosome Location
Chr. 6: 126708328 - 126743108 on Build GRCm38
UniGene ID
Mm.62535
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013568.6 NP_038596.1 1 2104
GenBank mRNA BC048782.1 - 1 1586
BC054804.1 - 1 1624 6838056
M96688.1 - 1 2269

View Full Product Details