Gene Transcripts

Gene Symbol
Mgrn1
Entrez Gene ID
17237
Gene Name
mahogunin, ring finger 1
Gene Aliases
2610042J20Rik, mKIAA0544, md, nc
Chromosome Location
Chr. 16: 4886100 - 4938296 on Build GRCm38
UniGene ID
Mm.291326
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252437.1 NP_001239366.1 4 695
NM_029657.4 NP_083933.1 4 692
GenBank mRNA AK011747.2 - 4 485
AK088533.1 - 4 475
AK129161.1 - 1 103
AK150176.1 - 5 528
AK153407.1 - 4 516
BC046830.1 - 4 541 6811606

View Full Product Details