Gene Transcripts

Gene Symbol
Evi5
Entrez Gene ID
14020
Gene Name
ecotropic viral integration site 5
Gene Aliases
NB4S
Chromosome Location
Chr. 5: 107744795 - 107875107 on Build GRCm38
UniGene ID
Mm.35796
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007964.2 NP_031990.2 18 3094
GenBank mRNA AK136326.1 - 8 1875
AK164208.1 - 4 1202
U53586.1 - 18 3093

View Full Product Details