Gene Transcripts

Gene Symbol
Fcna
Entrez Gene ID
14133
Gene Name
ficolin A
Gene Aliases
Fcn1
Chromosome Location
Chr. 2: 25624666 - 25635443 on Build GRCm38
UniGene ID
Mm.10510
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007995.3 NP_032021.1 8 659
GenBank mRNA AB007813.1 - 8 551
AB222271.1 - 7 440
AK133539.1 - 8 636
AK138752.1 - 8 606
AK159651.1 - 8 629
BC019180.1 - 8 607 5008934
CT010383.1 - 8 521

View Full Product Details