Gene Transcripts

Gene Symbol
Hiat1
Entrez Gene ID
15247
Gene Name
hippocampus abundant gene transcript 1
Gene Aliases
Chromosome Location
Chr. 3: 116631164 - 116662692 on Build GRCm38
UniGene ID
Mm.280077
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008246.2 NP_032272.2 12 2057
GenBank mRNA AK004779.1 - 12 1996
AK135865.1 - 12 1878
AK158838.1 - 12 2058
AK169626.1 - 12 1997
D88315.1 - 12 1930

View Full Product Details