Gene Transcripts

Gene Symbol
Il17ra
Entrez Gene ID
16172
Gene Name
interleukin 17 receptor A
Gene Aliases
AW538159, Cdw217, Il17r, VDw217
Chromosome Location
Chr. 6: 120463181 - 120483731 on Build GRCm38
UniGene ID
Mm.4481
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008359.2 NP_032385.1 3 398
GenBank mRNA AK036587.1 - 3 350
AK080226.2 - 3 349
U31993.1 - 3 386

View Full Product Details