Gene Transcripts

Gene Symbol
Ihh
Entrez Gene ID
16147
Gene Name
Indian hedgehog
Gene Aliases
Chromosome Location
Chr. 1: 74945318 - 74951651 on Build GRCm38
UniGene ID
Mm.439736
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010544.2 NP_034674.1 3 1677
GenBank mRNA AK144322.1 - 3 1700
BC046984.1 - 3 1677 6477118
U85610.1 - 3 1537

View Full Product Details