Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001042707.2 | NP_001036172.1 | 4 | 575 | |
NM_001042708.2 | NP_001036173.1 | 5 | 614 | ||
NM_001042709.2 | NP_001036174.1 | 4 | 575 | ||
NM_001277321.1 | NP_001264250.1 | 5 | 614 | ||
NM_001277322.1 | NP_001264251.1 | 6 | 542 | ||
NM_010561.3 | NP_034691.2 | 5 | 614 | ||
GenBank mRNA | AB221609.1 | - | 3 | 290 | |
AF098967.1 | - | 6 | 491 | ||
AF497751.2 | - | 5 | 482 | ||
AF497752.2 | - | 4 | 443 | ||
AK048096.1 | - | 4 | 505 | ||
AK077560.1 | - | 5 | 540 | ||
AK128936.1 | - | 5 | 547 | ||
AK150276.1 | - | 5 | 563 | ||
BC047272.1 | - | 5 | 534 | 6440312 | |
BC060599.1 | - | 3 | 278 | ||
DQ104405.1 | - | 5 | 482 | ||
DQ104406.1 | - | 4 | 443 |