Gene Transcripts

Gene Symbol
Gfpt1
Entrez Gene ID
14583
Gene Name
glutamine fructose-6-phosphate transaminase 1
Gene Aliases
2810423A18Rik, AI324119, AI449986, GFA, GFAT, GFAT1, GFAT1m, Gfpt
Chromosome Location
Chr. 6: 87042846 - 87092207 on Build GRCm38
UniGene ID
Mm.19893
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013528.3 NP_038556.1 11 1157
GenBank mRNA AF334736.1 - 12 1162
AK019852.1 - 11 1153
BC002283.1 - 8 756
BC010516.1 - 11 1105 4191553
BC050762.1 - 11 1153 6742987
U00932.1 - 10 1004

View Full Product Details