Gene Transcripts

Gene Symbol
Aqp7
Entrez Gene ID
11832
Gene Name
aquaporin 7
Gene Aliases
AQP7L, AQPap
Chromosome Location
Chr. 4: 41033074 - 41048215 on Build GRCm38
UniGene ID
Mm.8728
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007473.4 NP_031499.1 8 1570
GenBank mRNA AB056091.1 - 8 1404
BC022223.1 - 6 1200 4242417

View Full Product Details